Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005690417.2 | XP_005690474.1 | 4 - 5 | 492 | 77 | |
XM_018060102.1 | XP_017915591.1 | 4 - 5 | 474 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Capra hircus Probe spans exons |