Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018064668.1 | XP_017920157.1 | 4 - 5 | 829 | 71 | |
XM_018064669.1 | XP_017920158.1 | 4 - 5 | 981 | 71 | ||
XM_018064670.1 | XP_017920159.1 | 4 - 5 | 826 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Capra hircus Probe spans exons |