Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SDHAF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005699761.3 | XP_005699818.1 | 1 - 2 | 139 | 57 | |
GenBank mRNA | JI861816.1 | - | 1 - 2 | 34 | 57 | |
JO439119.2 | - | 1 - 2 | 137 | 57 | ||
JT754263.1 | - | 5 - 6 | 725 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Capra hircus Probe spans exons |