Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OPRD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018057063.1 | XP_017912552.1 | 2 - 3 | 826 | 102 | |
GenBank mRNA | JO437657.2 | - | 2 - 3 | 696 | 102 | |
JQ756319.1 | - | 1 - 2 | 208 | 102 | ||
JT672183.1 | - | 1 - 2 | 527 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Recommended Assay Capra hircus Probe spans exons |