Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SUSD5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018038257.1 | XP_017893746.1 | 3 - 4 | 392 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Capra hircus Probe spans exons |