Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018040209.1 | XP_017895698.1 | 4 - 5 | 451 | 90 | |
GenBank mRNA | JO580117.1 | - | 4 - 5 | 448 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Capra hircus Probe spans exons |