Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GAB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018043071.1 | XP_017898560.1 | 6 - 7 | 1656 | 67 | |
GenBank mRNA | JO578193.1 | - | 6 - 7 | 1609 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Capra hircus Probe spans exons |