Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKUB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018053811.1 | XP_017909300.1 | 2 - 3 | 350 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Capra hircus Probe spans exons |