Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLAMF8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005677242.3 | XP_005677299.1 | 4 - 5 | 996 | 94 | |
GenBank mRNA | JO585311.2 | - | 4 - 5 | 860 | 94 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Capra hircus Probe spans exons |