Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CFAP300 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005689373.3 | XP_005689430.2 | 4 - 5 | 467 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Capra hircus Probe spans exons |