Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KRT38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005693812.3 | XP_005693869.2 | 6 - 7 | 2494 | 68 | |
GenBank mRNA | KA342290.1 | - | 1 - 2 | 426 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Capra hircus Probe spans exons |