Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HESX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005695825.3 | XP_005695882.1 | 4 - 5 | 1752 | 83 | |
XM_005695826.3 | XP_005695883.1 | 3 - 4 | 1615 | 83 | ||
XM_018067186.1 | XP_017922675.1 | 5 - 6 | 749 | 83 | ||
XM_018067187.1 | XP_017922676.1 | 2 - 3 | 6372 | 83 | ||
GenBank mRNA | JT674056.1 | - | 6 - 7 | 1008 | 83 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Capra hircus Probe spans exons |