Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRRT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005697689.3 | XP_005697746.2 | 2 - 3 | 1779 | 91 | |
GenBank mRNA | JO645566.2 | - | 3 - 4 | 1265 | 91 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Capra hircus Probe spans exons |