Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC102187924 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005709716.3 | XP_005709773.2 | 2 | 1084 | 79 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Capra hircus Both primers and probe map within a single exon |