Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKFN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013971777.2 | XP_013827231.2 | 6 - 7 | 962 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Capra hircus Probe spans exons |