Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALKBH4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018040397.1 | XP_017895886.1 | 2 - 3 | 361 | 95 | |
GenBank mRNA | JO617216.2 | - | 2 - 3 | 382 | 95 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Capra hircus Probe spans exons |