Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCNK4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018043594.1 | XP_017899083.1 | 2 - 3 | 556 | 68 | |
GenBank mRNA | JO617886.2 | - | 2 - 3 | 457 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Capra hircus Probe spans exons |