Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: REG4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005677782.3 | XP_005677839.1 | 3 - 4 | 313 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Capra hircus Probe spans exons |