Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTPN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005679507.3 | XP_005679564.1 | 3 - 4 | 576 | 91 | |
GenBank mRNA | JO582116.1 | - | 1 - 2 | 1575 | 91 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Capra hircus Probe spans exons |