Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DCK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005681738.3 | XP_005681795.1 | 2 - 3 | 521 | 68 | |
GenBank mRNA | JO552186.2 | - | 1 - 2 | 197 | 68 | |
KA341652.1 | - | 2 - 3 | 440 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Capra hircus Probe spans exons |