Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPS14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005683172.3 | XP_005683229.1 | 4 - 5 | 644 | 66 | |
XM_005683173.3 | XP_005683230.1 | 4 - 5 | 625 | 66 | ||
GenBank mRNA | JO580974.1 | - | 4 - 5 | 689 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Capra hircus Probe spans exons |