Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM83C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005688673.3 | XP_005688730.3 | 3 - 4 | 991 | 102 | |
GenBank mRNA | KA330416.1 | - | 2 - 3 | 2031 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Capra hircus Probe spans exons |