Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRYAB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005689436.3 | XP_005689493.1 | 2 - 3 | 615 | 65 | |
GenBank mRNA | JO442347.2 | - | 2 - 3 | 625 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Capra hircus Probe spans exons |