Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDUFS4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005694716.2 | XP_005694773.1 | 2 - 3 | 200 | 72 | |
GenBank mRNA | JO516244.2 | - | 2 - 3 | 212 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Capra hircus Probe spans exons |