Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RUNDC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013972442.2 | XP_013827896.2 | 5 - 6 | 1057 | 60 | |
XM_018065157.1 | XP_017920646.1 | 4 - 5 | 1049 | 60 | ||
GenBank mRNA | JO628644.2 | - | 5 - 6 | 920 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Capra hircus Probe spans exons |