Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CASP7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005698497.3 | XP_005698554.1 | 6 - 7 | 818 | 62 | |
XM_018041622.1 | XP_017897111.1 | 6 - 7 | 702 | 62 | ||
GenBank mRNA | JI861721.1 | - | 5 - 6 | 687 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Capra hircus Probe spans exons |