Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NACC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018051240.1 | XP_017906729.1 | 2 - 3 | 1152 | 97 | |
GenBank mRNA | JO577993.1 | - | 4 - 5 | 1589 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Capra hircus Probe spans exons |