Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BCAR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018061715.1 | XP_017917204.1 | 2 - 3 | 2705 | 109 | |
XM_018061717.1 | XP_017917206.1 | 3 - 4 | 864 | 109 | ||
XM_018061718.1 | XP_017917207.1 | 3 - 4 | 897 | 109 | ||
XM_018061719.1 | XP_017917208.1 | 3 - 4 | 792 | 109 | ||
GenBank mRNA | JO637950.2 | - | 3 - 4 | 834 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Capra hircus Probe spans exons |