Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRPS1L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005679001.2 | XP_005679058.2 | 1 | 124 | 83 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Capra hircus Both primers and probe map within a single exon |