Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CXXC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005681344.3 | XP_005681401.3 | 1 - 2 | 1136 | 92 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Capra hircus Probe spans exons |