Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FGF17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005683972.2 | XP_005684029.1 | 3 - 4 | 418 | 75 | |
XM_005683973.2 | XP_005684030.1 | 3 - 4 | 385 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Capra hircus Probe spans exons |