Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XKRX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005700151.2 | XP_005700208.1 | 2 - 3 | 608 | 99 | |
GenBank mRNA | JO595045.2 | - | 2 - 3 | 365 | 99 | |
KA348053.1 | - | 2 - 3 | 1796 | 99 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Capra hircus Probe spans exons |