Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NKX2-8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013973096.2 | XP_013828550.2 | 1 - 2 | 381 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Capra hircus Probe spans exons |