Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VPS37D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018041070.1 | XP_017896559.1 | 2 - 3 | 424 | 106 | |
GenBank mRNA | JO642679.2 | - | 2 - 3 | 276 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Capra hircus Probe spans exons |