Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BCL7C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018040874.1 | XP_017896363.1 | 2 - 3 | 533 | 99 | |
XM_018040875.1 | XP_017896364.1 | 2 - 3 | 533 | 99 | ||
GenBank mRNA | JO408916.2 | - | 4 - 5 | 475 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Capra hircus Probe spans exons |