Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NEPRO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005674925.3 | XP_005674982.2 | 8 - 9 | 1305 | 100 | |
XM_013962566.2 | XP_013818020.2 | 7 - 8 | 1144 | 100 | ||
XM_018066177.1 | XP_017921666.1 | 8 - 9 | 1170 | 100 | ||
XM_018066237.1 | XP_017921726.1 | 8 - 9 | 1165 | 100 | ||
XM_018066416.1 | XP_017921905.1 | 5 - 6 | 627 | 100 | ||
GenBank mRNA | JO447536.2 | - | 1 - 2 | 696 | 100 | |
JO591728.2 | - | 1 - 2 | 641 | 100 | ||
KA336093.1 | - | 1 - 2 | 797 | 100 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Capra hircus Probe spans exons |