Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CALN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018041016.1 | XP_017896505.1 | 5 - 6 | 1314 | 86 | |
GenBank mRNA | JO645693.2 | - | 915 | 86 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Capra hircus Probe spans exons |