Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRC31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013964434.2 | XP_013819888.2 | 8 - 9 | 1267 | 90 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Capra hircus Probe spans exons |