Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLHL26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018051450.1 | XP_017906939.1 | 2 - 3 | 312 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Capra hircus Probe spans exons |