Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC102190399 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018051210.1 | XP_017906699.1 | 6 - 7 | 873 | 65 | |
GenBank mRNA | JT634544.1 | - | 7 - 8 | 971 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Capra hircus Probe spans exons |