Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NOVA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013971571.2 | XP_013827025.1 | 1 - 2 | 153 | 89 | |
GenBank mRNA | JO514999.2 | - | 1 - 2 | 130 | 89 | |
JO631216.2 | - | 866 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Capra hircus Probe spans exons |