Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DBF4B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018064012.1 | XP_017919501.1 | 5 - 6 | 564 | 87 | |
GenBank mRNA | JO582856.1 | - | 5 - 6 | 409 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Capra hircus Probe spans exons |