Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PNPLA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013973931.2 | XP_013829385.1 | 5 - 6 | 1292 | 83 | |
XM_018039266.1 | XP_017894755.1 | 5 - 6 | 1289 | 83 | ||
XM_018039267.1 | XP_017894756.1 | 5 - 6 | 1293 | 83 | ||
GenBank mRNA | KA327333.1 | - | 5 - 6 | 1330 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Capra hircus Probe spans exons |