Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD8B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018055352.1 | XP_017910841.1 | 2 - 3 | 452 | 85 | |
GenBank mRNA | JO326107.2 | - | 4 - 5 | 912 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Capra hircus Probe spans exons |