Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VPS37C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | KA311307.1 | - | 2 - 3 | 494 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Capra hircus Probe spans exons |