Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VPS37C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018043441.1 | XP_017898930.1 | 4 - 5 | 547 | 65 | |
GenBank mRNA | KA311307.1 | - | 1 - 2 | 407 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Capra hircus Probe spans exons |