Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NAALADL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005675296.3 | XP_005675353.2 | 12 - 13 | 2467 | 101 | |
XM_013964525.2 | XP_013819979.2 | 14 - 15 | 2221 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Capra hircus Probe spans exons |