Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NAALADL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005675296.3 | XP_005675353.2 | 7 - 8 | 1802 | 112 | |
XM_013964525.2 | XP_013819979.2 | 9 - 10 | 1556 | 112 | ||
GenBank mRNA | KA311265.1 | - | 3 - 4 | 457 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Capra hircus Probe spans exons |