Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TM4SF18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005675458.3 | XP_005675515.2 | 1 - 2 | 261 | 84 | |
XM_005675459.3 | XP_005675516.2 | 1 - 2 | 261 | 84 | ||
XM_018049095.1 | XP_017904584.1 | 1 - 2 | 221 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Capra hircus Probe spans exons |