Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NPVF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005679285.2 | XP_005679342.2 | 1 - 2 | 137 | 120 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Capra hircus Probe spans exons |